Learn More
Lenz Laborglasinstrumente™ Duran Tubing Reaction Vessels
Brand: Lenz Laborglasinstrumente™ 06000258
Additional Details : Weight : 1.00000kg
Specifications
500 mL | |
150 mm | |
Cylindrical |
Borosilicate Glass, Duran Glass | |
Reaction vessel | |
Flat |