Description
Hoses can be connected to laboratory apparatus via DURAN™ tube nozzles. There is always a risk of injury due to glass breakage when dismantling tightly fitted hoses from glass olives. DWK therefore recommends the use of DURAN™ glass threads with plastic nozzles.
- Hose olives enable the connection of hoses to laboratory apparatuses
- Glass articles for further processing
Specifications
Specifications
| Quantity | 250/Pk. |
| Item Description | For attaching to various apparatuses; Installation in different apparatus possible |
| Type | Tube with Connections |
| Joint Size | 6 mm |