Learn More
DWK Life Sciences DURAN™ Tube with Screw Thread, with DIN thread, and standard ground cone
Shop All DWK Life Sciences ProductsDescription
This threaded tube has a screw thread and a standard ground joint. Components of an apparatus can be connected via both ends and thus connected to each other.
- Tight closure due to standard ground joint
- With DIN thread
Specifications
Specifications
| Quantity | 10/Pk. |
| Item Description | Tube with screw threads |
| Type | Non-DIN Size |
| Standard Taper Joint No | GL 14, with Cone NS 24/29 |