Learn More
DWK Life Sciences DURAN™ Test Tube, with beaded rim or straight rim
Product Code. p-7189315 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
2 mL
4 mL
6 mL
8 mL
9 mL
16 mL
17 mL
21 mL
32 mL
34 mL
40 mL
55 mL
70 mL
100 mL
Unit Size:
Pack of 100
Pack of 50
Product Code. Capacity (Metric) unitSize
10508121 6 mL Pack of 100
10612781 16 mL Pack of 100
10716811 21 mL Pack of 100
10393411 32 mL Pack of 100
10403161 40 mL Pack of 100
10577731 2 mL Pack of 100
10445411 4 mL Pack of 100
10211721 17 mL Pack of 100
10038170 55 mL Pack of 50
10100551 70 mL Pack of 50
10068180 100 mL Pack of 50
10538311 8 mL Pack of 100
10493151 34 mL Pack of 100
11670029 9 mL Pack of 100
14 options
Product Code. 10508121
Brand: DWK Life Sciences 261310809
Only null left
Add to basket
Add to basket
Test tubes are universal cylindrical laboratory glassware with a round bottom that are used e.g. for chemical reactions, pretreatments or for storing small amounts of liquid.
Transferring substances and liquids into other states of aggregation and thus initiating further reactions is very important for the daily work in the laboratory. A wide variety of vessels, apparatus and equipment are used for these work. Reaction controls, test series and preliminary samples are usually carried out in test tubes due to the small amounts of substance removed.
  • Thick-walled and therefore mechanically very resistant
  • Available with beaded rim or straight rim
  • With round bottom
  • Fired-on, highly durable white ceramic
TRUSTED_SUSTAINABILITY
Specifications
Description 0.8 to 1.0 mm thickness
Capacity (Metric) 6 mL
Material Borosilicate Glass (3.3)
For Use With Heating of small amounts; Pretreatments; Storage and transport of small amounts of liquid
Quantity 100/Pk.
Rimmed Straight Rim
Autoclavable Autoclavable
Diameter (Metric) 12 mm