Learn More
DWK Life Sciences DURAN™ Standard Socket, double, diamond-ground
Shop All DWK Life Sciences Products
missing translation for 'orderingAttributeHoverText'
Joint Size:
18 mm
28 mm
34 mm
missing translation for 'unitSize'
Each
Pack of 50
Description
DURAN™ DG double sleeves are not ground according to DIN 12249 and represent the cost-effective alternative to DURAN™ cores.
- Diamond cut (coarser cut quality, more roughness depth)
- Cost-effective alternative to DURAN™ sleeves (not according to DIN)
- Glass article with two sleeves enables material savings
- Tight closure due to standard ground joint
- Glass articles for further processing
Specifications
Specifications
| Quantity | 50/Pk. |
| Item Description | Installation in glass apparatus and devices |
| Joint Size | 18 mm |
| Thickness (Metric) Wall | 1.8 mm |