Learn More
DWK Life Sciences DURAN™ Standard Cone, double, finished
Product Code. p-7189324 Shop All DWK Life Sciences Products
Click to view available options
Joint Size:
13 mm
17 mm
22 mm
26 mm
Unit Size:
Pack of 25
Pack of 50
Product Code. 15844598
Brand: DWK Life Sciences 286172809
Only null left
Add to basket
Add to basket
For a rigid, self-supporting connection.
The DURAN™ double cores, ground, are an old, less common type of design according to DIN 12249 Form D. Their primary purpose is to save material.
  • According to DIN 12249 Form D
  • Ground (finer grinding quality, less roughness depth)
  • Glass article with two cores enables material savings
  • Tight closure due to standard ground joint
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Quantity 50/Pk.
Item Description Installation in glass apparatus and devices
Joint Size 13 mm
Thickness (Metric) Wall 1.5 mm