Learn More
DWK Life Sciences DURAN™ Single way Stopcock, complete with PTFE key
Shut-off with PTFE plug for grease free operation.
146.17 NOK - 12780.00 NOK
Specifications
Material | Borosilicate Glass (3.3), PTFE |
---|---|
Autoclavable | Yes |
For Use With (Application) | Installation in glass apparatus and devices |
Product Code | Brand | Closure Size | Plug Bore | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Size | Plug Bore | Price | Quantity & Availability | ||||
15884718
|
DWK Life Sciences
286414601 |
21.5 NS | 6 mm |
2700.00 NOK
Pack of 5 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
15844718
|
DWK Life Sciences
286412806 |
12.5 NS | 2.5 mm |
10695.00 NOK
Pack of 50 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
15804728
|
DWK Life Sciences
286415006 |
29.2 NS | 10 mm |
4915.00 NOK
Pack of 5 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
15854718
|
DWK Life Sciences
286413605 |
14.5 NS | 4 mm |
12780.00 NOK
Pack of 50 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
15864718
|
DWK Life Sciences
286413802 |
18.8 NS | 4 mm |
8235.00 NOK
Pack of 25 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
15874718
|
DWK Life Sciences
286414207 |
18.8 NS | 6 mm |
8740.00 NOK
Pack of 25 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
15894718
|
DWK Life Sciences
286414807 |
24 NS | 8 mm |
3605.00 NOK
Pack of 5 |
Estimated Shipment: 20-06-2024
Log in to see stock. |
||||
15834718
|
DWK Life Sciences
286412609 |
12.5 NS | 2 mm |
10695.00 NOK
Pack of 50 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
10263781
|
DWK Life Sciences
286413408 |
14.5 NS | 2.5 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
DURAN™ single way stopcocks complete with PTFE valve plugs allow grease free operation. The 3-part tap safety catch secures the tap plug in the tap sleeve.
- Polished shut-off valve sleeve with PTFE valve plug for grease-free operation
- The 3-part tap safety catch secures the tap plug in the tap sleeve.
- Taper 1:10
- Glass articles for further processing
Specifications
Borosilicate Glass (3.3), PTFE | |
Yes | |
Installation in glass apparatus and devices |