Learn More
DWK Life Sciences DURAN™ Single way Stopcock, complete with PTFE key
Product Code. 15894718 Shop All DWK Life Sciences Products
Change view
Click to view available options
Plug Bore:
2 mm
2.5 mm
4 mm
6 mm
8 mm
10 mm
Closure Size:
12.5 NS
14.5 NS
18.8 NS
21.5 NS
24 NS
29.2 NS
Unit Size:
Pack of 25
Pack of 5
Pack of 50
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Bore Closure Size Unit Size
15894718 8 mm 24 NS Pack of 5
15884718 6 mm 21.5 NS Pack of 5
15844718 2.5 mm 12.5 NS Pack of 50
15804728 10 mm 29.2 NS Pack of 5
15854718 4 mm 14.5 NS Pack of 50
15864718 4 mm 18.8 NS Pack of 25
15874718 6 mm 18.8 NS Pack of 25
15834718 2 mm 12.5 NS Pack of 50
10263781 2.5 mm 14.5 NS Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 15894718 Supplier DWK Life Sciences Supplier No. 286414807
  • ONLINE EXCLUSIVE PRICE
    4055.00 NOK
    3277.04 NOK / Pack of 5
    Save 777.96 NOK
    19% Off
Estimated Shipment: 21-12-2026
Shut-off with PTFE plug for grease free operation.
DURAN™ single way stopcocks complete with PTFE valve plugs allow grease free operation. The 3-part tap safety catch secures the tap plug in the tap sleeve.
  • Polished shut-off valve sleeve with PTFE valve plug for grease-free operation
  • The 3-part tap safety catch secures the tap plug in the tap sleeve.
  • Taper 1:10
  • Glass articles for further processing
Specifications
Plug Bore 8 mm
Diameter (Metric) 15 mm
Material Borosilicate Glass (3.3), PTFE
Autoclavable Autoclavable
Closure Size 24 NS
Width (Metric) 1.8 mm
No. per Pack 5/Pk.
For Use With (Application) Installation in glass apparatus and devices