Learn More
DWK Life Sciences DURAN™ Round Bottom Flask, wide neck, with beaded rim
Product Code. p-7189278 Shop All DWK Life Sciences Products
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
2000 mL
4000 mL
6000 mL
Unit Size:
Each
Pack of 10
Product Code. 11736093
Brand: DWK Life Sciences 217412407
Only null left
Add to basket
Add to basket
Due to their spherical shape, round bottom flasks are used for heating liquids, among other things.
Abdominal vessels are called flasks in the laboratory and, in addition to beakers, are other universally applicable vessels. They are used e.g. for distillation or for heating liquids. Flasks are available in various designs: narrow and wide neck, with or without ground joint, as round or flat bottom flask. Depending on the application, the required flask with its specific components is selected.
  • The wide neck enables easy filling and cleaning
  • The geometry permits very uniform heating
  • Fired-on, highly durable white ceramic
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Base 64 mm
Capacity (Metric) 100 mL
Material Borosilicate Glass (3.3)
For Use With (Application) Suitable for heating mantles; Heating; Mixing several components; Sizes 50, 1000, 2000, 3000, 5000, 6000, 10000, 12000 and 20000 mL non-DIN ISO size
No. per Pack 10/Pk.