Learn More
DWK Life Sciences™ DURAN™ Replacement Handle and Collar, for GL 45 Bottle Carrier
Product Code. p-7189428 Shop All DWK Life Sciences Products
Click to view available options
For Use With (Equipment):
For GL 45 5 L Bottle Carrier
Unit Size:
Each
Product Code. 16474954
Brand: DWK Life Sciences™ 292847304
Only null left
Add to basket
Add to basket
For easy and safe carrying and pouring of GL 45 laboratory glass bottles.
The new modular Bottle Carrying System fits standard laboratory bottles that have a GL 45 neck finish and is compatible with four bottle sizes: 2, 5, 10 and 20 liters. It is ergonomically designed, and offers health and safety benefits to the user and allows easy and safe transport of bottles around the laboratory or production facility. It makes pouring liquids from the larger bottles more comfortable and safer. All materials are fully autoclavable. The collar is only removable by cutting – so attachment is 100 % secure in use.
  • The ergonomical design offers health and safety benefits and allows easy and safe transport of bottles
  • It makes pouring liquids from larger bottles more comfortable and safer
  • The collar is only removable by cutting what makes the attachment 100 % secure in use
  • Supplied without glass bottle
TRUSTED_SUSTAINABILITY
Specifications
Product Type Replacement Bottle Carrier
For Use With (Equipment) For GL 45 5 L Bottle Carrier