Learn More
DWK Life Sciences DURAN™ Reduction Adapter, socket/cone
Product Code. 10223881 Shop All DWK Life Sciences Products
Change view
Click to view available options
Joint Size:
Socket NS 19/26, Cone NS 14/23
Socket NS 24/29, Cone NS 14/23
Socket NS 24/29, Cone NS 19/26
Socket NS 29/32, Cone NS 14/23
Socket NS 29/32, Cone NS 19/26
Socket NS 29/32, Cone NS 24/29
Unit Size:
Each
Pack of 10
Product Code. Joint Size unitSize
10223881 Socket NS 29/32, Cone NS 24/29 Each
15884818 Socket NS 29/32, Cone NS 14/23 Pack of 10
10305101 Socket NS 29/32, Cone NS 19/26 Each
15981601 Socket NS 24/29, Cone NS 19/26 Pack of 10
15961601 Socket NS 19/26, Cone NS 14/23 Pack of 10
15844818 Socket NS 24/29, Cone NS 14/23 Pack of 10
6 options
Product Code. 10223881
Brand: DWK Life Sciences 286548201
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Reduction from sleeve to core.
Adapters are used to connect two components with different NS sizes. DURAN™ reducers allow the connection of a larger sleeve to a smaller core.
  • Reducers allow the connection of a larger sleeve to a smaller core and thus the use of differently sized ground joint components.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10/Pk.
Item Description Connection of two components with different standard ground joint sizes
Joint Size Socket NS 29/32, Cone NS 24/29