Description
In addition to solid-liquid filtration, filtration or separation of reaction mixtures from the gas phase is also possible. If, for example, a pipeline filter is installed in a hose system, these filters solid residues from the gas phase and thus purifies it.
- By installing the filter in a pipe system, the purification of a carrier gas is possible
- Fired-on, highly durable white print
Specifications
Specifications
| Material | Borosilicate Glass (3.3) |
| For Use With (Application) | In-line filtration of gas lines to remove solid impurities (e.g. dust); Qualitative inorganic analysis and preparative chemistry; Filtration at reduced pressure; Filtering |
| No. per Pack | 1/Pk. |
| Porosity | 3 |
| Diameter (Metric) | 90 mm |