Learn More
DWK Life Sciences DURAN™ Pipeline Filter
A pipeline filter is used to separate solids from a gas phase.
Brand: DWK Life Sciences 258550104
684.00 NOK valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 0.02600kg
Description
In addition to solid-liquid filtration, filtration or separation of reaction mixtures from the gas phase is also possible. If, for example, a pipeline filter is installed in a hose system, these filters solid residues from the gas phase and thus purifies it.
- By installing the filter in a pipe system, the purification of a carrier gas is possible
- Fired-on, highly durable white print
Specifications
Porosity 1 | |
In-line filtration of gas lines to remove solid impurities (e.g. dust); Qualitative inorganic analysis and preparative chemistry; Filtration at reduced pressure; Filtering | |
30 mm |
Borosilicate Glass (3.3) | |
1/Pk. |