Learn More
DWK Life Sciences DURAN™ Multi-Connector Cap, GL 45, Tubing Connectors, for peristaltic tubing, 2, 3, 4, or 5 tubing connectors, Red PBT screw cap
Shop All DWK Life Sciences ProductsDescription
The reusable DURAN™ GL 45 Connection Cap System is ideal for aseptic fluid transfer into or out of DURAN™ bottles using peristaltic pump tubing. The screw caps will fit all DURAN™ Original GL 45 laboratory bottles 100 mL and larger. The barbed tubing connectors (4.2 mm outer diameter) accept narrow BPT peristaltic pump tubing with a 3.2 mm (1/8) inner diameter.
- Caps have a GL 45 thread that fit DURAN™ bottles
- Barbed male hose connector are suitable for attaching peristaltic PBT tubing with a 4.2 mm inner diameter
- Available with 2, 3, 4, or 5 stainless steel (316L) barbed tubing connectors
- Autoclavable at 121° C or 134°C
- Construction materials are certified as approved for food contact
- Reusable with a long service life
Specifications
Specifications
| For Use With (Application) | Media supply for small bioreactors; Fed-batch bioreactor; Perfusion circuit for cell culture; Oxygenation/aeration of very small samples; Use in a chemistry micro-reactor |
| Product Type | Connector Cap (4-port) |