Learn More
DWK Life Sciences DURAN™ Mixing Cylinder, with Hexagonal Base, Class A
Product Code. 15230856 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Pack of 2
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15230856 10 mL Pack of 2
15240856 25 mL Pack of 2
15250856 50 mL Pack of 2
15260856 100 mL Pack of 2
15270856 250 mL Pack of 2
15280856 500 mL Pack of 2
15290856 1000 mL Each
15200866 2000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 15230856 Supplier DWK Life Sciences Supplier No. 246180856
Trusted Glassware Deals
Use promo code 28509
View all promotions
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
DURAN™ Mixing cylinders are for diluting solutions and mixing several components in a given quantity ratio.
DURAN™ Mixing cylinders are for diluting solutions and mixing several components in a given quantity ratio. Available in two different accuracy Classes – Class A with main ring points graduation and Class B with line graduation. For additional differentiation have the mixing cylinders with accuracy Class A a blue graduation and the B Class mixing cylinders a white graduation. Volume content tolerances for measuring and mixing cylinders conform to DIN and ISO accuracy limits. Thus the desired concentration can be precisely set. Mixing cylinders with a batch number and accuracy Class A are supplied with a batch certificate. This certificate documents the mean value obtained from measuring the batch in question, the standard deviation and the day of issue. The batch certificates can also be retrieved online. The batch number consists of four digits. The first two numbers specify the production year, and the following two numbers specify the batch, e.g.: 18.01.
  • Calibration based on the poured in volume (In) at a + 20°C reference temperature
  • A uniform wall thickness over the entire measuring range prevents wedge errors
  • Large hexagonal base equipped with three knobs which increase its stability and prevent the cylinder from rolling
  • Supplied with ergonomic polyethylene stopper for easy and safe tightening or removal
  • Tight closure due to standard ground joint
  • Fired-on, highly durable blue print
  • Easy-to-read scale with graduation marks to estimate the content
  • With circular ring graduations
  • The volume content tolerances conform to accuracy Class A
TRUSTED_SUSTAINABILITY
Specifications
Tolerance ± 0.1 mL
Material Borosilicate Glass (3.3), Polyethylene
Capacity (Metric) 10 mL
For Use With (Application) Measuring liquid amounts; Diluting solutions; Mixing several components; Volumetric measurement; To ensure a long service life for your volumetric glassware and to exclude possible volume changes, these glass products should not be heated above + 250°C in drying cabinets or sterilisers
Includes 2/Pk.
Autoclavable Autoclavable
Diameter (Metric) Outer 14 mm