Learn More
DWK Life Sciences DURAN™ Microbiology Bottle Unground Wide Neck
Tous les produits DWK Life Sciences ProduitsDescription
The culture bottles are moulded from DURAN™ borosilicate 3.3 Type 1 glass, and are suitable for the preparation of media or culturing of microorganisms. The bottles comply with ISO 4796-2 and feature a wide conical unground neck for use with rubber stoppers. Suitable for autoclaving at 121 or 134°C or dry heat sterilization.
- Wide neck for rubber stoppers
- Wide neck for easier filling and cleaning
- Retrace code
- Fired-on, highly durable white print
- Autoclavable at 121° C or 134°C
- Supplied without stopper
Specifications
Specifications
| Capacity (Metric) | 1000 mL |
| For Use With (Application) | Autoclaving of media and reagents; Preparation of media and reagents; Culturing vessel |
| Material | Borosilicate Glass (3.3) |
| Quantity | 10/Pk. |
| Diameter (Metric) | 107 mm |
| Autoclavable | Autoclavable |