Learn More
Lenz Laborglasinstrumente™ Duran™ Medium-neck Round Bottom Flasks
Product Code. 10220831
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Diameter (Metric) Outer:
31 mm
45 mm
51 mm
64 mm
85 mm
105 mm
131 mm
166 mm
Standard Taper Joint:
NS 14/23
NS 29/32
Unit Size:
Each
Pack of 10
Product Code. Capacity (Metric) Diameter (Metric) Outer Standard Taper Joint unitSize
10220831 500 mL 105 mm NS 29/32 Each
10483861 1000 mL 131 mm NS 29/32 Pack of 10
15360895 10 mL 31 mm NS 14/23 Pack of 10
10559421 100 mL 64 mm NS 29/32 Each
11737425 100 mL 64 mm NS 29/32 Pack of 10
10589611 2000 mL 166 mm NS 29/32 Each
10211551 250 mL 85 mm NS 29/32 Pack of 10
15370885 25 mL 45 mm NS 14/23 Pack of 10
11915956 500 mL 105 mm NS 29/32 Pack of 10
10710992 50 mL 51 mm NS 14/23 Pack of 10
10508851 50 mL 51 mm NS 29/32 Each
11 options
Product Code. 10220831
Brand: Lenz Laborglasinstrumente™ 03002958
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured from Duran™ glass. Lenz Laborglasinstrumente™ Duran™ Medium-neck Round Bottom Flasks include a conical ground joint.
  • Manufactured from Duran™ glass
  • Complies with DIN EN ISO 4797 standards
  • Includes conical ground joint
Specifications
Certifications/Compliance DIN EN ISO 4797
Capacity (Metric) 500 mL
Material Borosilicate Glass, Duran Glass
Neck Style Medium
Standard Taper Joint NS 29/32
Bottom Shape Round
Diameter (Metric) Outer 105 mm