Learn More
DWK Life Sciences DURAN™ Long Ground Socket, finished
Product Code. 15834828 Shop All DWK Life Sciences Products
Click to view available options
Joint Size:
17 mm
22 mm
28 mm
33 mm
Unit Size:
Pack of 50
Product Code. 15834828
Brand: DWK Life Sciences 286558408
Only null left
Add to basket
Add to basket
For ground joints in the high vacuum range.
DURAN™ sleeves in the long-grinding variant are frequently used in the high-vacuum range.
  • Ground (finer grinding quality, less roughness depth)
  • Short grinding for apertures with low space availability and lower tightness requirements
  • Tight closure due to standard ground joint
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Quantity 50/Pk.
Item Description Use in the high vacuum range; Installation in glass apparatus and devices
Joint Size 33 mm
Thickness (Metric) Wall 2 mm