Learn More
DWK Life Sciences DURAN™ Long Ground Cone with tip, finished
Product Code. 15834588 Shop All DWK Life Sciences Products
Change view
Click to view available options
Joint Size:
13 mm
22 mm
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Joint Size unitSize
15834588 13 mm Pack of 50
15854588 22 mm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15834588 Supplier DWK Life Sciences Supplier No. 286164809
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For ground joints in the high-vacuum range with extension.
The DURAN™ long ground cores with outlet combine the advantages of long ground and outlet cores.
  • Ground (finer grinding quality, less roughness depth)
  • The larger ground surface is suitable for use in high-vacuum applications.
  • With drain
  • Tight closure due to standard ground joint
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Quantity 50/Pk.
Item Description The additional outlet can also be used as an attachment point for extensions for various purposes, e.g. as an inlet pipe; Installation in glass apparatus and devices
Joint Size 13 mm
Standard Taper Joint No NS 14/35
Thickness (Metric) Wall 1.5 mm