Learn More
DWK Life Sciences DURAN™ Long Ground Cone, finished
Product Code. 15834568 Shop All DWK Life Sciences Products
Change view
Click to view available options
Joint Size:
13 mm
17 mm
22 mm
26 mm
40 mm
Unit Size:
Pack of 10
Pack of 50
Product Code. Joint Size unitSize
15834568 17 mm Pack of 50
15884568 26 mm Pack of 50
15864568 22 mm Pack of 50
15884578 40 mm Pack of 10
15894558 13 mm Pack of 50
5 options
Product Code. 15834568
Brand: DWK Life Sciences 286155804
Only null left
Add to basket
Add to basket
For ground joints in the high vacuum range.
DURAN™ cores in the long-grinding variant are frequently used in the high-vacuum range.
  • Ground (finer grinding quality, less roughness depth)
  • The larger ground surface is suitable for use in high-vacuum applications.
  • Tight closure due to standard ground joint
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Quantity 50/Pk.
Item Description Use in the high vacuum range; Installation in glass apparatus and devices
Joint Size 17 mm
Thickness (Metric) Wall 1.8 mm