Learn More
DWK Life Sciences DURAN™ KPG™ Stirrer Bearing, interchangeable, HB
Product Code. p-7189270 Shop All DWK Life Sciences Products
Change view
Click to view available options
Joint Size:
10 mm
16 mm
Unit Size:
Each
Product Code. Joint Size unitSize
10517351 16 mm Each
10748531 10 mm Each
2 options
Product Code. 10517351
Brand: DWK Life Sciences 245065708
Only null left
Add to basket
Add to basket
KPG bearing sleeve is a precisely fitting holder for guiding the KPG agitator shaft.
KPG agitators consist of a KPG agitator shaft and a KPG bearing sleeve. KPG bearing parts are manufactured with high accuracy and uniformity. The specified tolerances for nominal diameter and fit are maintained by careful checks. This guarantees the interchangeability of the KPG bearing sleeves and KPG stirrer shafts. Due to the low bearing clearance between the running surfaces, almost absolute sealing and completely quiet running can be achieved with suitable lubrication. KPG agitators are an alternative to magnetic stirrers and are used for reaction vessels that are not accessible from below when very viscous reaction mixtures are to be stirred or a high stirring speed is required.
  • High accuracy and uniformity within defined tolerances for nominal diameter and fit
  • Ground and polished bearing surface
  • Interchangeable
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Ea.
Item Description Stirring of liquids (for syntheses); Stirring at high stirring speed; Mounting support
Joint Size 16 mm