Learn More
DWK Life Sciences DURAN™ HPLC Reservoir Bottle, Clear, Conical Base, GL 45
GREENER_CHOICE
Product Code. 15819007 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
10,000 mL
1000 mL
2000 mL
3500 mL
5000 mL
Packungsgröße:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Capacity (Metric) unitSize
15819007 1000 mL Each
15829007 2000 mL Each
15839007 5000 mL Each
15849007 10,000 mL Each
16444944 3500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 15819007 Lieferant DWK Life Sciences Lieferanten-Nr. 218415407
Spotlight Offers
Use promo code 28036
Alle Werbeaktionen anzeigen
In Stock
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
The optimized conical design allows the safest and most efficient delivery of mobile phase to HPLC analysers.
The DURAN™ reservoir bottles have been specially designed for the storage and delivery of high-performance liquid chromatography (HPLC) mobile phases and solvents. Reservoirs have a one central cavity for the inlet filter to allow the most efficient delivery of mobile phase to HPLC analysers. A safer and more convenient alternative to flat bottomed glass media bottles, the conical bottom reservoirs allow a greater recovery of solvent, without having to tilt reservoir and risk a solvent spill. The larger reservoirs allow longer, uninterrupted analyses. These are especially useful during a weekend or lengthy chromatographic analysis when there might be a risk of running out of solvents during unattended operation.
  • The optimized conical design allows the safest and most efficient delivery of mobile phase to HPLC analysers
  • Manufactured from the material of choice for solvent bottles: borosilicate 3.3 glass. This material has an exceptional chemical durability offering a very low inorganic ion extractables profile, and the total absence of any organic extractables.
  • DURAN™ is a borosilicate glass type 3.3 as specified by the international standard ISO 3585, and is classified as a Type I, Class A low-expansion borosilicate glass as defined by ASTM-E438-1992
  • DURAN™ glass is classified by the USP / EP / JP pharmacopoeias as a Type I neutral glass
  • Retrace Code, and certificate for manufacturing lot traceability
  • On the two larger sizes, the central cavity can accommodate magnetic stir bars for improved mixing
  • Side vent / drain hole in the base for ultrasonic bath degassing, or water drainage after cleaning
  • GL 45 thread fits standard HPLC connection caps
  • Durable white enamel volume graduations
TRUSTED_SUSTAINABILITY
Spezifikation
Capacity (Metric) 1000 mL
Material Borosilicate Glass (3.3)
For Use With (Application) Supply of solvents to HPLC equipment; Safe transfer of liquid media within a closed and sterile system
Includes Threads, size GL 45
No. per Pack 1/Pk.
Autoclavable Autoclavable
Diameter (Metric) Outer 110 mm