Learn More
DWK Life Sciences DURAN™ Hexagonal Stopper, pointed, diamond-ground
Product Code. 15854548 Shop All DWK Life Sciences Products
Click to view available options
Size:
NS 10/19
NS 12/21
NS 14/23
NS 19/26
NS 24/29
NS 29/32
NS 34/35
NS 45/40
NS 7/16
Unit Size:
Each
Pack of 10
Pack of 100
Product Code. 15854548
Brand: DWK Life Sciences 286113808
Only null left
Add to basket
Add to basket
Hollow glass stopper for sealing with draining tip.
DURAN™ hexagonal hollow plugs are used for sealing laboratory equipment such as volumetric flasks, mixing cylinders, ground joint flasks, etc. DURAN™ hexagonal hollow plugs are available in 2 standard versions - pointed - flat - Sizes analogous to DURAN™ conical ground joints. Cut quality: DG = Diamond Ground
  • Pointed
  • Diamond-ground
  • With a dripping syringe, for example, a condensate can be collected and the reaction medium recirculated.
  • hollow glass stopper
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Closure of vessels with standard ground joints
Quantity 100/Pk.
Size NS 7/16
Autoclavable Autoclavable
Material Borosilicate Glass
Product Content Correction
Product Title
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.