Learn More
Lenz Laborglasinstrumente™ Duran Glass Straight Gas Inlet Tubes
Product Code. 10342851
Change view
Click to view available options
Length (Metric):
200mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10342851 200mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10342851 Supplier Lenz Laborglasinstrumente™ Supplier No. 05080001
  • ONLINE EXCLUSIVE PRICE
    272.00 NOK
    129.13 NOK / Pack of 1
    Save 142.87 NOK
    52% Off
Estimated Shipment: 04-11-2026
Specifications
Includes NS cone and thin capillary
Length (Metric) 200mm
Type Gas Inlet Tube
Material Duran Glass
Quantity 1Each
Standard Taper Joint NS 14/23