Learn More
Lenz Laborglasinstrumente™ DURAN™ Glass Stirrer
Click to view available options
For Use With (Equipment):
Flask, 10,000 mL
Flask, 100 mL
Flask, 1000 mL
Flask, 2000 mL
Flask, 250 mL
Flask, 250/500 mL
Flask, 250/500 mL, 10 GL Thread
Flask, 4000/5000 mL
Unit Size:
Each
Description
- Duran™ Glass
Specifications
Specifications
| For Use With (Equipment) | Flask, 250/500 mL, 10 GL Thread |
| Product Type | Stirrer |
| Material | Glass |