Learn More
Lenz Laborglasinstrumente™ Duran™ Glass Spiral Condensers
Product Code. 10633891
Change view
Click to view available options
Length (Metric) Jacket:
160 mm
250 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Jacket unitSize
10633891 250 mm Each
10111571 160 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10633891 Supplier Lenz Laborglasinstrumente™ Supplier No. 05233314
  • ONLINE EXCLUSIVE PRICE
    1360.00 NOK
    914.69 NOK / Pack of 1
    Save 445.31 NOK
    32% Off
Estimated Shipment: 19-08-2026
Specifications
For Use With (Application) For cooling condensates
Length (Metric) Jacket 250 mm
Quantity 1 Each
Standard Taper Joint No 29/32
Material Duran Glass
Type Spiral Condenser