Learn More
Lenz Laborglasinstrumente™ Screwthread Tubes with NS Cone
Description
- Acc. to DIN 12257
- Available in a range of socket and thread sizes
- Minimized need for grease
- Corrosion-free and resistant to thermal shock
- With conical joint, featuring high taper accuracy and excellent tightness
Specifications
Specifications
| Quantity | 1 Ea. |
| Cone Size | NS 29/31 |
| Standard Taper Joint No | GL 14 |
| Material | Duran Glass |