Learn More
Lenz Laborglasinstrumente™ Reduction Adapters with Ground Joints
Click to view available options
Cone Size:
NS 24/29
Unit Size:
Each
Description
- Acc. to DIN 12257
- High taper accuracy of the cones
- Minimized need for grease
- Excellent tightness
- Corrosion-free and resistant to thermal shock
Specifications
Specifications
| Certifications/Compliance | DIN 12257 and in addition to DIN standard |
| Cone Size | NS 24/29 |
| Material | Duran Glass |
| Product Type | Reduction Adapter |
| Quantity | 1Each |