Learn More
Lenz Laborglasinstrumente™ Duran™ Glass GL 14 Thread Dimroth Condensers
Product Code. 11516814
Change view
Click to view available options
Standard Taper Joint No:
14/23
19/26
29/32
Length (Metric) Jacket:
160 mm
250 mm
400 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Length (Metric) Jacket Unit Size
11516814 19/26 250 mm Each
10550003 14/23 160 mm Each
11596804 19/26 160 mm Each
10762903 29/32 400 mm Each
10030182 29/32 250 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11516814 Supplier Lenz Laborglasinstrumente™ Supplier No. 05240312
  • ONLINE EXCLUSIVE PRICE
    1545.00 NOK
    1279.67 NOK / Pack of 1
    Save 265.33 NOK
    17% Off
Estimated Shipment: 04-11-2026
Specifications
Certifications/Compliance DIN 12591 and in addition to DIN standard
Description Cooling Surface: 440cm2
For Use With (Application) For condensing vapors
Length (Metric) Jacket 250 mm
Quantity 1 Each
Standard Taper Joint No 19/26
Material Duran Glass
Type Dimroth Condenser