Learn More
Lenz Laborglasinstrumente™ Expansion Adapter with Ground Joint
Description
- Acc. to DIN 12257
- High taper accuracy of the cones
- Minimized need for grease
- Excellent tightness
- Corrosion-free and resistant to thermal shock
Specifications
Specifications
| Certifications/Compliance | DIN 12257 and in addition to DIN standard |
| Cone Size | NS 19/26 |
| Material | Duran Glass |
| Product Type | Expansion Adapter |
| Quantity | 1 Ea. |
| Standard Taper Joint No. | NS 24/29 |