Learn More
Lenz Laborglasinstrumente™ Expansion Adapter with Ground Joint
Product Code. 10742682
Change view
Click to view available options
Cone Size:
NS 14/23
NS 19/26
NS 24/29
NS 29/32
Standard Taper Joint No.:
NS 19/26
NS 24/29
NS 29/32
NS 45/40
Unit Size:
Each
Product Code. Cone Size Standard Taper Joint No. unitSize
10742682 NS 14/23 NS 19/26 Each
10529951 NS 24/29 NS 19/26 Each
12694873 NS 19/26 NS 24/29 Each
10312711 NS 19/26 NS 29/32 Each
10323331 NS 29/32 NS 45/40 Each
10149740 NS 14/23 NS 29/32 Each
6 options
Product Code. 10742682
Brand: Lenz Laborglasinstrumente™ 01501914
Only null left
Add to basket
Add to basket
Provides for secure and easy connection from a small cone to a large socket in glass tubing. Lenz Laborglasinstrumente™ Expansion Adapters with Ground Joints are manufactured from durable DURAN™ borosilicate glass.
  • Acc. to DIN 12257
  • High taper accuracy of the cones
  • Minimized need for grease
  • Excellent tightness
  • Corrosion-free and resistant to thermal shock
Specifications
Certifications/Compliance DIN 12257 and in addition to DIN standard
Cone Size NS 14/23
Material Duran Glass
Product Type Expansion Adapter
Quantity 1 Ea.
Standard Taper Joint No. NS 19/26