Learn More
Lenz Laborglasinstrumente™ Duran™ Glass Dimroth Condensers
Product Code. 11866194
Change view
Click to view available options
Standard Taper Joint No:
19/26
29/32
Length (Metric) Jacket:
160 mm
250 mm
400 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Length (Metric) Jacket Unit Size
11866194 29/32 250 mm Each
10642282 19/26 160 mm Each
10642472 19/26 250 mm Each
10017810 29/32 400 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11866194 Supplier Lenz Laborglasinstrumente™ Supplier No. 05240304
  • ONLINE EXCLUSIVE PRICE
    1465.00 NOK
    1214.59 NOK / Pack of 1
    Save 250.41 NOK
    17% Off
Estimated Shipment: 18-12-2026
Specifications
Certifications/Compliance DIN 12591 and in addition to DIN standard
Description Cooling Surface: 440cm2
For Use With (Application) For condensing vapors
Length (Metric) Jacket 250 mm
Quantity 1 Each
Standard Taper Joint No 29/32
Material Duran Glass
Type Dimroth Condenser