Learn More
Lenz Laborglasinstrumente™ Duran™ Glass Dimroth Condensers
Product Code. 10642282
Change view
Click to view available options
Standard Taper Joint No:
19/26
29/32
Length (Metric) Jacket:
160 mm
250 mm
400 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Length (Metric) Jacket Unit Size
10642282 19/26 160 mm Each
10642472 19/26 250 mm Each
11866194 29/32 250 mm Each
10017810 29/32 400 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10642282 Supplier Lenz Laborglasinstrumente™ Supplier No. 05240202
  • ONLINE EXCLUSIVE PRICE
    1170.00 NOK
    969.04 NOK / Pack of 1
    Save 200.96 NOK
    17% Off
Estimated Shipment: 07-01-2027
Specifications
Certifications/Compliance acc. to DIN 12591 and in addition to DIN standard
Description Cooling Surface: 140cm2
For Use With (Application) For condensing vapors
Length (Metric) Jacket 160 mm
Quantity 1 Each
Standard Taper Joint No 19/26
Material Duran Glass
Type Dimroth Condenser