Learn More
Lenz Laborglasinstrumente™ Duran™ Glass Dimroth Condensers
Product Code. 10017810
Change view
Click to view available options
Standard Taper Joint No:
19/26
29/32
Length (Metric) Jacket:
160 mm
250 mm
400 mm
Unit Size:
Each
Product Code. Standard Taper Joint No Length (Metric) Jacket unitSize
10017810 29/32 400 mm Each
10642282 19/26 160 mm Each
10642472 19/26 250 mm Each
11866194 29/32 250 mm Each
4 options
Product Code. 10017810
Brand: Lenz Laborglasinstrumente™ 05240404
Only null left
Specifications
Certifications/Compliance DIN 12591 and in addition to DIN standard
Description Cooling Surface: 740cm2
For Use With (Application) For condensing vapors
Length (Metric) Jacket 400 mm
Quantity 1 Each
Standard Taper Joint No 29/32
Material Duran Glass
Type Dimroth Condenser