Learn More
Lenz Laborglasinstrumente™ Duran™ Gas Sampling Tube
Product Code. 10796981
Change view
Click to view available options
Capacity (Metric):
150 mL
350 mL
500 mL
1000 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10796981 350 mL Each
10737181 1000 mL Each
10221851 150 mL Each
10057970 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10796981 Supplier Lenz Laborglasinstrumente™ Supplier No. 05680152
  • ONLINE EXCLUSIVE PRICE
    1385.00 NOK
    931.73 NOK / Pack of 1
    Save 453.27 NOK
    32% Off
View all promotions
Estimated Shipment: 11-01-2027
Specifications
Capacity (Metric) 350 mL
Material Duran Glass
Type Gas Sampling Tube
Includes Nozzle GL14 Thread and septa
Certifications/Compliance DIN 12473-1
Quantity 1 Pack