Description
- Soft flexible EPDM rubber forms an air-tight seal
- EPDM ensures an airtight seal with bottle
Specifications
Specifications
| Description | 40 mm ring gasket seal for GL45 multi-function caps. Circular EPDM Seals 1.5 mm thick with a 40 mm outer and 29 mm inner diameter. |
| For Use With (Application) | Sterile fluid transfer in and out of lab bottles; Gas transfer in and out of lab bottles; Fluid transfer bewteen lab bottles; Supply of reagents to instruments |
| Joint Size | GL 45 |
| Quantity | 5/Pk. |
| Material | EPDM |