Learn More
Lenz Laborglasinstrumente™ Duran™ Flat Flange Lids
Specifications
Specifications
| Material | Borosilicate Glass, Duran Glass |
| For Use With (Equipment) | Reaction Vessels |
| Height (Metric) | 125mm |
| No. per Pack | 1 |
| Product Type | Flat Flange Lid |
| Closure Type | Flange Lid |