Learn More
DWK Life Sciences DURAN™ Flat Flange Lid, 4 standard ground necks, with side necks (standard ground): 3 x 29/32 angled, Center (standard ground): 29/32
Product Code. 10190151 Shop All DWK Life Sciences Products
Change view
Click to view available options
Description:
DN 100
DN 150
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
10190151 DN 150 Each
10088240 DN 100 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10190151 Supplier DWK Life Sciences Supplier No. 243925801
  • ONLINE EXCLUSIVE PRICE
    4140.00 NOK
    3345.19 NOK / Pack of 1
    Save 794.81 NOK
    19% Off
Estimated Shipment: 03-12-2026
The lid of a plane fan vessel is suitable for closing but also for combination with other ground joint components (e.g. dropping funnel, stirring shafts, etc.) due to the standard ground joints.
Flat flange vessels are large volume reaction vessels which are regarded as an alternative to the round flasks and consist of a bottom part and a lid. These two parts are held together by means of a clamping ring. A seal or grinding grease prevent the grinding surfaces from tilting. The flat flange vessel can be combined with other components (e.g. dropping funnels, stirring shafts, ...) by means of various standard ground joints in the lid. Flat flange vessels are well suited for preparative work such as general reactions, distillations, evaporation and drying of solid and viscous substances. Due to their walls, flat flange vessels are suitable for safe working under pressure and vacuum.
  • With one middle neck: 29/32 and three side necks (standard ground): 3 x 29/32 parallel
  • Pressure and vacuum resistant due to the wall thickness and geometry
  • Tight closure due to standard ground joint
Specifications
Description DN 150
Material Borosilicate Glass (3.3)
For Use With (Application) Chemically and thermally demanding processes; Safe working under pressure or vacuum
Neck Style 3 x NS 29/32 angled
No. per Pack 1/Pk.
Socket Joint NS 29/32