Learn More
DWK Life Sciences DURAN™ Flat Flange Lid, 4 standard ground necks, with side necks: 2 x NS 29/32 angled; 1 x 14/23 parallel,, center neck: NS 29/32
Shop All DWK Life Sciences ProductsDescription
Flat flange vessels are large volume reaction vessels which are regarded as an alternative to the round flasks and consist of a bottom part and a lid. These two parts are held together by means of a clamping ring. A seal or grinding grease prevent the grinding surfaces from tilting. The flat flange vessel can be combined with other components (e.g. dropping funnels, stirring shafts, ...) by means of various standard ground joints in the lid. Flat flange vessels are well suited for preparative work such as general reactions, distillations, evaporation and drying of solid and viscous substances. Due to their walls, flat flange vessels are suitable for safe working under pressure and vacuum.
- With one middle neck: 29/32 and three side necks (standard ground): 3 x 29/32 parallel
- Pressure and vacuum resistant due to the wall thickness and geometry
- Tight closure due to standard ground joint
Specifications
Specifications
| Description | DN 100 |
| Material | Borosilicate Glass (3.3) |
| For Use With (Application) | Chemically and thermally demanding processes; Safe working under pressure or vacuum |
| Neck Style | 2 x NS 29/32 angled; 1 x NS 14/23 parallel |
| No. per Pack | 1/Pk. |
| Socket Joint | NS 29/32 |
| Autoclavable | Autoclavable |