Learn More
DWK Life Sciences DURAN™ Flat Flange, ground
Shop All DWK Life Sciences Products
Click to view available options
Diameter (Metric) Base:
70 mm
106 mm
130 mm
155 mm
215 mm
Includes:
With groove
Without groove
Unit Size:
Pack of 2
Description
DURAN™ flat flanges with and without grooves are suitable for further processing into pressure-resistant reaction vessels, for example. For pressure resistance, please refer to our notes in the technical appendix of the laboratory glassware catalogue. For glass articles which deviate from the original form due to further processing, the permissible operating data must be determined in accordance with the technical rules and regulations.
- Ground
- Component for further processing into reaction vessels
- Due to the geometry and the massive wall thickness, the blanks are suitable for devices with pressure and vacuum applications.
- Glass articles for further processing
Specifications
Specifications
| Diameter (Metric) Base | 106 mm |
| Material | Borosilicate Glass (3.3) |
| For Use With (Application) | For processing as a reaction vessel |
| Includes | With groove |
| No. per Pack | 2/Pk. |
| Autoclavable | Autoclavable |