Learn More
DWK Life Sciences DURAN™ Flat Bottom Flask, wide neck, with beaded rim
Shop All DWK Life Sciences Products
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Pack of 10
Description
Abdominal vessels are called flasks in the laboratory and, in addition to beakers, are other universally applicable vessels. They are used e.g. for distillation or for heating liquids. Flasks are available in various designs: narrow and wide neck, with or without ground joint, as round or flat bottom flask. Depending on the application, the required flask with its specific components is selected.
- Flat base means flasks can be set down without a supporting ring
- The wide neck enables easy filling and cleaning
- Fired-on, highly durable white ceramic
Specifications
Specifications
| Diameter (Metric) Base | 51 mm |
| Capacity (Metric) | 50 mL |
| Material | Borosilicate Glass (3.3) |
| For Use With (Application) | Heating; Size 2000 mL non-DIN EN ISO size |
| No. per Pack | 10/Pk. |
| Autoclavable | Autoclavable |