Learn More
Lenz Laborglasinstrumente™ Extractor Heads Acc. to Soxhlet
Product Code. 10392951
Change view
Click to view available options
Capacity (Metric):
30 mL
70 mL
100 mL
150 mL
250 mL
500 mL
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10392951 150 mL Each
10740612 100 mL Each
10251741 250 mL Each
10700612 30 mL Each
10662361 70 mL Each
10343051 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10392951 Supplier Lenz Laborglasinstrumente™ Supplier No. 05395143
  • ONLINE EXCLUSIVE PRICE
    1290.00 NOK
    897.21 NOK / Pack of 1
    Save 392.79 NOK
    30% Off
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
For the extraction of liquids with specific light solvents. Lenz Laborglasinstrumente™ Extractor Heads Acc. to Soxhlet are manufactured from durable DURAN™ tubing.
  • Acc to DIN 12602 and Soxhlet
  • With cone NS 29/32
  • Available with/without NS stopcock
  • Corrosion-free and resistant to thermal shock
Specifications
Product Type Extractor Head
Capacity (Metric) 150 mL
Material Duran Glass