Learn More
Lenz Laborglasinstrumente™ Extractor Heads Acc. to Soxhlet
Product Code. 10343051
Click to view available options
Capacity (Metric):
30 mL
70 mL
100 mL
150 mL
250 mL
500 mL
Unit Size:
Each
Product Code. 10343051
Brand: Lenz Laborglasinstrumente™ 05395158
For the extraction of liquids with specific light solvents. Lenz Laborglasinstrumente™ Extractor Heads Acc. to Soxhlet are manufactured from durable DURAN™ tubing.
  • Acc to DIN 12602 and Soxhlet
  • With cone NS 29/32
  • Available with/without NS stopcock
  • Corrosion-free and resistant to thermal shock
Specifications
Product Type Extractor Head
Capacity (Metric) 500 mL
Material Duran Glass
Certifications/Compliance DIN 12602