Learn More
Karl Hecht™ DURAN™ Extraction Apparatus with Dimroth Condenser
Product Code. 10257041
Change view
Click to view available options
Standard Taper Joint Flask:
29/32
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint Flask Unit Size
10257041 29/32 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10257041 Supplier Karl Hecht™ Supplier No. 41151012
  • ONLINE EXCLUSIVE PRICE
    2910.00 NOK
    1726.97 NOK / Pack of 1
    Save 1183.03 NOK
    40% Off
 Restricted Item
Estimated Shipment: 30-12-2026
This extraction apparatus is according to Soxhlet.
  • With standard grounds
  • Flask joint size 29/32, condenser joint size of 34/35
  • Joint sizes and capacity of extractors are durably marked
  • Consists of extractor, dimroth condenser and round bottom flask
  • Flask capacity of 100mL, extraction capacity of 70mL
Specifications
Product Type Soxhlet Extraction Apparatus
Capacity (Metric) 100 mL (Flask), 70 mL (Extractor)
Material Borosilicate Glass 3.3
Standard Taper Joint Condenser 34/35
Standard Taper Joint Flask 29/32