Learn More
DWK Life Sciences DURAN™ Expansion Adapter, socket/cone
Product Code. 15834808 Shop All DWK Life Sciences Products
Change view
Click to view available options
Joint Size:
Socket NS 14/23, Cone NS 19/26
Socket NS 14/23, Cone NS 24/29
Socket NS 14/23, Cone NS 29/32
Socket NS 19/26, Cone NS 24/29
Socket NS 19/26, Cone NS 29/32
Socket NS 24/29, Cone NS 29/32
Socket NS 24/29, Cone NS 45/40
Socket NS 29/32, Cone NS 34/35
Socket NS 29/32, Cone NS 45/40
Socket NS 29/32, Cone NS 50/42
Socket NS 29/32, Cone NS 60/46
Socket NS 29/32, Cone NS 71/51
Unit Size:
Each
Pack of 10
Product Code. Joint Size unitSize
15834808 Socket NS 14/23, Cone NS 24/29 Pack of 10
15931601 Socket NS 14/23, Cone NS 19/26 Pack of 10
10142021 Socket NS 24/29, Cone NS 45/40 Each
15941601 Socket NS 14/23, Cone NS 29/32 Pack of 10
15884808 Socket NS 19/26, Cone NS 24/29 Pack of 10
15894808 Socket NS 19/26, Cone NS 29/32 Pack of 10
15931611 Socket NS 29/32, Cone NS 45/40 Pack of 10
15804828 Socket NS 29/32, Cone NS 60/46 Pack of 10
15814828 Socket NS 29/32, Cone NS 71/51 Pack of 10
10161781 Socket NS 24/29, Cone NS 29/32 Each
15921611 Socket NS 29/32, Cone NS 34/35 Pack of 10
10707101 Socket NS 29/32, Cone NS 34/35 Each
15894818 Socket NS 29/32, Cone NS 50/42 Pack of 10
13 options
Product Code. 15834808
Brand: DWK Life Sciences 286524609
Only null left
Add to basket
Add to basket
Expansion from sleeve to core.
Adapters are used to connect two components with different NS sizes. DURAN™ expansion pieces allow the connection of a smaller sleeve to a larger core.
  • Expansion pieces allow the connection of a smaller sleeve to a larger core and thus the use of differently sized ground joint components.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10/Pk.
Item Description Connection of two components with different standard ground joint sizes
Joint Size Socket NS 14/23, Cone NS 24/29