Learn More
DWK Life Sciences DURAN™ Erlenmeyer Flask, with standard ground joint
Shop All DWK Life Sciences ProductsDescription
Due to their large footprint, Erlenmeyer flasks are suitable, for example, for preparing solutions and storing them. The additional grinding allows a tight closure of the vessel.
- Tight closure due to standard ground joint
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white print
- Easy mixing due to conical form
Specifications
Specifications
| Diameter (Metric) Base | 105 mm |
| Capacity (Metric) | 500 mL |
| Material | Borosilicate Glass (3.3) |
| For Use With (Application) | Mixing several components; Diluting solutions; Measuring liquid amounts; Heating; Volume sizes 50 mL (NS 24/29), 100 mL (NS 24/29), 200 mL (NS 29/32), 250 mL (NS 45/40), 300 mL (NS 29/32), 500 mL (NS 45/40) and 1000 mL (NS 45/40) are not according to DIN ISO |
| No. per Pack | 10/Pk. |
| Product Type | Non-DIN/ISO size |
| Thread Size | NS 45/40 |
| Autoclavable | Autoclavable |