Learn More
DWK Life Sciences DURAN™ Dropping Funnel, cylindrical, with scale
Product Code. 10233361 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10233361 100 mL Each
10060131 500 mL Each
10408582 250 mL Each
10439922 50 mL Each
10101551 1000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10233361 Supplier DWK Life Sciences Supplier No. 241222409
Trusted Glassware Deals
Use promo code 28509
View all promotions
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
The dropping funnel allows a dosed addition of solutions.
The dosed addition of solutions, reaction mixtures and/or chemicals is ideally carried out using a dropping funnel.
  • With standard ground stopcock and retaining device
  • The rate of supply can be adjusted
  • Cylindrical with scale
  • Tight closure due to standard ground joint
  • Fired-on, highly durable white print
  • Easy-to-read scale with graduation marks to estimate the content
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 9 mm
Capacity (Metric) 100 mL
Material Boro 3.3, Polypropylene
Autoclavable Autoclavable
For Use With (Application) Uniform and metered liquid supply to a reaction mixture; Measuring liquid amounts
Graduations 2 mL
Quantity 1 Ea.