Learn More
Hirschmann™ DURAN™ Dafert Pattern Burettes with Blue Graduations
Excellent for dispensing measured amounts of chemical solutions in the laboratory.
2925.00 NOK - 2990.00 NOK
Specifications
Description | Burettes, acc. to Dafert, with three-way stopcock and glass key, with filling funnel |
---|---|
Material | Glass |
Includes | Schellbach Stripe |
Scale | Blue |
Stopcock | Straight |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
13198009
|
Hirschmann™
3330170 |
25 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
13108019
|
Hirschmann™
3330175 |
50 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Schellbach
- Short line
- With blue graduation
- Three-way stopcock with glass key
- Straight form
Specifications
Burettes, acc. to Dafert, with three-way stopcock and glass key, with filling funnel | |
Schellbach Stripe | |
Straight | |
Dafert Pattern Buret |
Glass | |
Blue | |
Glass |