Learn More
DWK Life Sciences DURAN™ Culture Flask, Erlenmeyer shape, straight neck for metal caps
Product Code. 11712919 Shop All DWK Life Sciences Products
Click to view available options
Capacity (Metric):
100 mL
200 mL
250 mL
300 mL
500 mL
1000 mL
2000 mL
Unit Size:
Pack of 10
Product Code. 11712919
Brand: DWK Life Sciences 217713209
Only null left
Add to basket
Add to basket
Culture bottles are suitable for attaching culture media.
Microbiology is an important field of biology and medicine and deals with microorganisms (bacteria, fungi, protozoa or viruses) and their habitats and conditions. For this purpose, it is essential to produce pure culture media on which the corresponding microorganisms can grow. Irradiation of the media with IR or UV light can accelerate the growth of the cultures. Culture bottles have large bottom surfaces that are as flat as possible so that the layer thickness of the medium is uniformly high. The straight necks of the bottles allow Kapsenberg and metal caps to be inserted.
  • Due to their conical geometry, these flasks are very suitable for shake tests (e.g. media optimization)
  • The straight neck permits the use of metal caps
  • Easy-to-read scale with graduation marks to estimate the content
  • Fired-on, highly durable white print
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Base 79 mm
Capacity (Metric) 200 mL
Material Borosilicate Glass (3.3)
For Use With (Application) Media preparation and cultivation; Preparation of cultures; Liquid culture; Mixing several components; Measuring liquid amounts
No. per Pack 10/Pk.