Learn More
DWK Life Sciences DURAN™ Crystallizing Dish, without spout
Product Code. 11786582 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
20 mL
40 mL
60 mL
100 mL
150 mL
300 mL
500 mL
900 mL
3500 mL
Unit Size:
Pack of 10
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11786582 150 mL Pack of 10
11746582 20 mL Pack of 10
11756582 40 mL Pack of 10
11766582 60 mL Pack of 10
11776582 100 mL Pack of 10
11746592 300 mL Pack of 10
11756592 500 mL Pack of 10
11766592 900 mL Pack of 10
15221689 3500 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 11786582 Supplier DWK Life Sciences Supplier No. 213134104
  • ONLINE EXCLUSIVE PRICE
    1410.00 NOK
    1263.07 NOK / Pack of 10
    Save 146.93 NOK
    10% Off
Estimated Shipment: 21-12-2026
The flat shape as well as the flat bottom of the crystallizing dish allow an even evaporation of solvents and thus the crystallization of solids from solutions and suspensions.
Transferring substances and liquids into other states of aggregation and thus initiating further reactions is very important for the daily work in the laboratory. A wide variety of vessels, apparatus and equipment are used for these work.
  • Flat and wide shape allow solvents to evaporate quickly and evenly
  • Large ground base
  • Spout for clean pouring
  • Fired-on, highly durable white ceramic
Specifications
Autoclavable Autoclavable
Capacity (Metric) 150 mL
Material Borosilicate Glass (3.3)
For Use With (Application) Crystallization; Preparation of concentrated solutions; Available with spout DIN 12 338 or without spout DIN 12 337
No. per Pack 10/Pk.
Diameter (Metric) 80 mm